accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3C(Z)

CAS RN 928-55-2

IUPAC Name: (1Z)-1-ethoxyprop-1-ene
Synonyms: (1Z)-1-ethoxyprop-1-ene
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: XDHOEHJVXXTEDV-HYXAFXHYSA-N
SMILES: CCO\C=C/C
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (2)