accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 928-89-2

IUPAC Name: 6-chlorohex-1-ene
Synonyms: 6-chlorohex-1-ene
Molecular Weight (g/mol): 118.6
Molecular Formula: C6H11Cl
InChi Key: BLMIXWDJHNJWDT-UHFFFAOYSA-N
SMILES: ClCCCCC=C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)