accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 928-94-9

IUPAC Name: (2Z)-hex-2-en-1-ol
Synonyms: (2Z)-hex-2-en-1-ol
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: ZCHHRLHTBGRGOT-PLNGDYQASA-N
SMILES: CCC\C=C/CO
Boiling Point 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)