accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 928038-44-2

IUPAC Name: (azetidin-3-yl)methanol hydrochloride
Synonyms: azetidin-3-ylmethanol hydrochloride
Molecular Weight (g/mol): 123.58
Molecular Formula: C4H10ClNO
InChi Key: AQUVQGSNKVDBBF-UHFFFAOYSA-N
SMILES: Cl.OCC1CNC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results