CAS RN 928207-02-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ ErbB2 Inhibitor II
The ErbB2 Inhibitor II, also referenced under CAS 928207-02-7, controls the biological activity of ErbB2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.