accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH

CAS RN 92866-00-7

IUPAC Name: 1-ethynyl-4-(2-phenylethynyl)benzene
Synonyms: 1-ethynyl-4-(2-phenylethynyl)benzene
Molecular Weight (g/mol): 202.26
Molecular Formula: C16H10
InChi Key: WAUKYPQSFIENDE-UHFFFAOYSA-N
SMILES: C#CC1=CC=C(C=C1)C#CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results