accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBrF

CAS RN 928715-37-1

IUPAC Name: 1-(2-bromo-6-fluorophenyl)ethan-1-one
Synonyms: 1-(2-bromo-6-fluorophenyl)ethanone
Molecular Weight (g/mol): 217.04
Molecular Formula: C8H6BrFO
InChi Key: ZTLLHQUYOLPVAR-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results