accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 928783-85-1

IUPAC Name: 1-bromo-3-chloro-5-(trifluoromethyl)benzene
Synonyms: 1-bromo-3-chloro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 259.45
Molecular Formula: C7H3BrClF3
InChi Key: QRPRXRQFNNXPBA-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results