accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 928830-73-3

IUPAC Name: 1-(5-fluoro-2,4-dinitrophenyl)-4-methylpiperazine
Synonyms: 1-(5-fluoro-2,4-dinitrophenyl)-4-methylpiperazine
Molecular Weight (g/mol): 284.25
Molecular Formula: C11H13FN4O4
InChi Key: SSBAFRIGYXRBRK-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results