accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92976-81-3

IUPAC Name: N,N-dimethylpyridin-4-amine; tribroman-2-uide
Synonyms: DMAP; tribroman-2-uide
Molecular Weight (g/mol): 361.88
Molecular Formula: C7H10Br3N2
InChi Key: TYDZHQMKSLAFKM-UHFFFAOYSA-N
SMILES: Br[Br-]Br.CN(C)C1=CC=NC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)