accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOH3CCH3NOH3CCH3CH3CH3OHOCH3OHOH2ORh

CAS RN 929896-28-6

IUPAC Name: 2-{2,4-dimethyl-5-[4-(propan-2-yl)-4,5-dihydro-1,3-oxazol-2-yl]phenyl}-4-(propan-2-yl)-4,5-dihydro-1,3-oxazole bis(acetic acid) hydrate rhodium
Synonyms: 4-isopropyl-2-[5-(4-isopropyl-4,5-dihydro-1,3-oxazol-2-yl)-2,4-dimethylphenyl]-4,5-dihydro-1,3-oxazole bis(acetic acid) hydrate rhodium
Molecular Weight (g/mol): 569.48
Molecular Formula: C24H38N2O7Rh
InChi Key: PRWDPLSLZWMIPQ-UHFFFAOYNA-N
SMILES: O.[Rh].CC(O)=O.CC(O)=O.CC(C)C1COC(=N1)C1=CC(C2=NC(CO2)C(C)C)=C(C)C=C1C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results