accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93-02-7

IUPAC Name: 2,5-dimethoxybenzaldehyde
Synonyms: 2,5-dimethoxybenzaldehyde
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: AFUKNJHPZAVHGQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(C=O)=C1
Boiling Point 
  • (5)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (4)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)