accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 93-09-4

IUPAC Name: naphthalene-2-carboxylic acid
Synonyms: β-naphthoic acid isonaphthoic acid 2-maythic acid ne-2-carboxylic acid
Molecular Weight (g/mol): 172.18
Molecular Formula: C11H8O2
InChi Key: UOBYKYZJUGYBDK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)