accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH2OCH3

CAS RN 93-15-2

IUPAC Name: 1,2-dimethoxy-4-(prop-2-en-1-yl)benzene
Synonyms: methyl eugenol methyl eugenyl ether 1,3,4-eugenol methyl ether fema number 2475 caswell no. 579AB eugenyl methyl ether ccris 746 3,4-dimethoxyallylbenzene epa pesticide chemical code 203900
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: ZYEMGPIYFIJGTP-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC=C)C=C1OC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)