accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93-17-4

IUPAC Name: 2-(3,4-dimethoxyphenyl)acetonitrile
Synonyms: benzylcyanide,3,4-dimethoxy
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: ASLSUMISAQDOOB-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC#N)C=C1OC
Boiling Point 
  • (2)
  • (1)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (5)
  • (2)
  • (4)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)