accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93-40-3

IUPAC Name: 2-(3,4-dimethoxyphenyl)acetic acid
Synonyms: 3,4-dimethoxyphenylacetic acid
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: WUAXWQRULBZETB-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(O)=O)C=C1OC
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (1)
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (6)
  • (4)
  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)