accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNH

CAS RN 93-46-9

IUPAC Name: N1,N4-bis(naphthalen-2-yl)benzene-1,4-diamine
Synonyms: diafen NN
Molecular Weight (g/mol): 360.46
Molecular Formula: C26H20N2
InChi Key: VETPHHXZEJAYOB-UHFFFAOYSA-N
SMILES: N(C1=CC=C(NC2=CC=C3C=CC=CC3=C2)C=C1)C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results