accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93-68-5

IUPAC Name: N-(2-methylphenyl)-3-oxobutanamide
Synonyms: N-(2-methylphenyl)-3-oxobutanamide
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: TVZIWRMELPWPPR-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC=CC=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results