accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNCl

CAS RN 93-70-9

IUPAC Name: N-(2-chlorophenyl)-3-oxobutanamide
Synonyms: N-(2-chlorophenyl)-3-oxobutanamide
Molecular Weight (g/mol): 211.65
Molecular Formula: C10H10ClNO2
InChi Key: BFVHBHKMLIBQNN-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC=CC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results