accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOClClCl

CAS RN 93-76-5

IUPAC Name: 2-(2,4,5-trichlorophenoxy)acetic acid
Synonyms: fortex VEON caswell no. 881 envert-T trichlorophenoxyacetic acid decamine 4T forron farmco fence rider epa pesticide chemical code 082001 arbokan
Molecular Weight (g/mol): 255.48
Molecular Formula: C8H5Cl3O3
InChi Key: SMYMJHWAQXWPDB-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results