accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 93-91-4

IUPAC Name: 1-phenylbutane-1,3-dione
Synonyms: 1,3-butanedione, 1-phenyl-
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: CVBUKMMMRLOKQR-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (8)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (3)
  • (2)
  • (3)
  • (6)
  • (6)
  • (8)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)