accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH2

CAS RN 930-02-9

IUPAC Name: 1-(ethenyloxy)octadecane
Synonyms: polyvinyl stearyl ether
Molecular Weight (g/mol): 296.54
Molecular Formula: C20H40O
InChi Key: QJJDJWUCRAPCOL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results