accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)(R)H3C

CAS RN 930-90-5

IUPAC Name: (1R,2R)-1-ethyl-2-methylcyclopentane
Synonyms: (1R,2R)-1-ethyl-2-methylcyclopentane
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: BSKOLJVTLRLTHE-HTQZYQBOSA-N
SMILES: CC[C@@H]1CCC[C@H]1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results