accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONHSH3COHO

CAS RN 93000-03-4

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-(methylsulfanyl)butanoic acid|BOC-DL-MET-OH
Molecular Weight (g/mol): 249.33
Molecular Formula: C10H19NO4S
InChi Key: IMUSLIHRIYOHEV-UHFFFAOYNA-N
SMILES: CSCCC(NC(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)