accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 930110-98-8

IUPAC Name: [5-(pyrrolidin-1-yl)pyridin-2-yl]methanol
Synonyms: [5-(pyrrolidin-1-yl)pyridin-2-yl]methanol
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: GLWULQJNDPJFDR-UHFFFAOYSA-N
SMILES: OCC1=NC=C(C=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results