accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 930110-99-9

IUPAC Name: 5-(furan-2-yl)pyridine-2-carboxylic acid
Synonyms: 5-(furan-2-yl)pyridine-2-carboxylic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: LTWJYKGQIACFMN-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=C(C=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results