accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNNH3C

CAS RN 930111-04-9

IUPAC Name: methyl[(1-methyl-1H-pyrazol-5-yl)methyl]amine
Synonyms: methyl[(2-methylpyrazol-3-yl)methyl]amine
Molecular Weight (g/mol): 125.18
Molecular Formula: C6H11N3
InChi Key: CHGIPIYKGPNYKT-UHFFFAOYSA-N
SMILES: CNCC1=CC=NN1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results