accessibility menu, dialog, popup

UserName

Viewing profile as user:
SBrOOH

CAS RN 930111-09-4

IUPAC Name: 4-(3-bromothiophen-2-yl)benzoic acid
Synonyms: 4-(3-bromothiophen-2-yl)benzoic acid
Molecular Weight (g/mol): 283.14
Molecular Formula: C11H7BrO2S
InChi Key: WAOAWGITQLHUFM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=C(Br)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results