accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONOOH3CNOO

CAS RN 930111-86-7

IUPAC Name: methyl 1-(4-methylbenzenesulfonyl)-4-nitro-1H-pyrrole-2-carboxylate
Synonyms: methyl 1-(4-methylbenzenesulfonyl)-4-nitropyrrole-2-carboxylate
Molecular Weight (g/mol): 324.31
Molecular Formula: C13H12N2O6S
InChi Key: AYFBDJWRDWDJJZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CN1S(=O)(=O)C1=CC=C(C)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results