accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOO

CAS RN 931-40-8

IUPAC Name: 4-(hydroxymethyl)-1,3-dioxolan-2-one
Synonyms: hydroxymethyl dioxolanone
Molecular Weight (g/mol): 118.09
Molecular Formula: C4H6O4
InChi Key: JFMGYULNQJPJCY-UHFFFAOYNA-N
SMILES: OCC1COC(=O)O1
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)