accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHMgBr

CAS RN 931-50-0

IUPAC Name: magnesium(2+) cyclohexanide bromide
Synonyms: magnesium(2+) cyclohexanide bromide
Molecular Weight (g/mol): 187.36
Molecular Formula: C6H11BrMg
InChi Key: GFTXWCQFWLOXAT-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].C1CC[CH-]CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results