accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 931-61-3

IUPAC Name: N-methylpyrimidin-2-amine
Synonyms: N-methylpyrimidin-2-amine
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: BQNXHDSGGRTFNX-UHFFFAOYSA-N
SMILES: CNC1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results