accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOH

CAS RN 931-71-5

IUPAC Name: cyclohexane-1,4-diol
Synonyms: 1,4-cyclohexanediol, trans-
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: VKONPUDBRVKQLM-UHFFFAOYSA-N
SMILES: OC1CCC(O)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results