accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 931-88-4

IUPAC Name: (Z)-cyclooctene
Synonyms: cyclooctene
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: URYYVOIYTNXXBN-UPHRSURJSA-N
SMILES: C1CCC\C=C/CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results