accessibility menu, dialog, popup

UserName

Viewing profile as user:
MgBrCH3

CAS RN 932-31-0

IUPAC Name: bromo(2-methylphenyl)magnesium
Synonyms: bromo(2-methylphenyl)magnesium
Molecular Weight (g/mol): 195.34
Molecular Formula: C7H7BrMg
InChi Key: DVXDIGKNJYSMFM-UHFFFAOYSA-M
SMILES: CC1=CC=CC=C1[Mg]Br
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)