accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 932-66-1

IUPAC Name: 1-(cyclohex-1-en-1-yl)ethan-1-one
Synonyms: 1-(cyclohex-1-en-1-yl)ethanone
Molecular Weight (g/mol): 124.18
Molecular Formula: C8H12O
InChi Key: LTYLUDGDHUEBGX-UHFFFAOYSA-N
SMILES: CC(=O)C1=CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)