accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSO

CAS RN 932-95-6

IUPAC Name: thiophene-2,5-dicarbaldehyde
Synonyms: thiophene-2,5-dicarbaldehyde
Molecular Weight (g/mol): 140.16
Molecular Formula: C6H4O2S
InChi Key: OTMRXENQDSQACG-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(S1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results