accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93224-85-2

IUPAC Name: 2-bromo-6-chlorobenzoic acid
Synonyms: 2-bromo-6-chlorobenzoic acid
Molecular Weight (g/mol): 235.46
Molecular Formula: C7H4BrClO2
InChi Key: URGXUQODOUMRFP-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results