accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 932739-76-9

IUPAC Name: N2,N7-bis(naphthalen-1-yl)-N2,N7-diphenyl-9,9'-spirobi[fluorene]-2,7-diamine
Synonyms: N2,N7-bis(naphthalen-1-yl)-N2,N7-diphenyl-9,9'-spirobi[fluorene]-2,7-diamine
Molecular Weight (g/mol): 750.95
Molecular Formula: C57H38N2
InChi Key: ZDAWFMCVTXSZTC-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC2=C(C=C1)C1=C(C=C(C=C1)N(C1=CC=CC=C1)C1=C3C=CC=CC3=CC=C1)C21C2=CC=CC=C2C2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results