CAS RN 932749-62-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Histone Acetyltransferase Inhibitor II
The Histone Acetyltransferase Inhibitor II, also referenced under CAS 932749-62-7, controls the biological activity of Histone Acetyltransferase. This small molecule/inhibitor is primarily used for Cell Structure applications.