accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 932750-29-3

IUPAC Name: ethyl 2-cyano-3-ethoxybut-2-enoate
Synonyms: ethyl 2-cyano-3-ethoxybut-2-enoate
Molecular Weight (g/mol): 183.21
Molecular Formula: C9H13NO3
InChi Key: YKXFOGAYPIPTKF-UHFFFAOYSA-N
SMILES: CCOC(C)=C(C#N)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results