accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3

CAS RN 933-40-4

IUPAC Name: 1,1-dimethoxycyclohexane
Synonyms: 1,1-dimethoxycyclohexane
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: XPIJMQVLTXAGME-UHFFFAOYSA-N
SMILES: COC1(CCCCC1)OC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)