accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 933-75-5

IUPAC Name: 2,3,6-trichlorophenol
Synonyms: 2,3,6-trichlorophenol
Molecular Weight (g/mol): 197.44
Molecular Formula: C6H3Cl3O
InChi Key: XGCHAIDDPMFRLJ-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=CC(Cl)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results