accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CH3C

CAS RN 933-98-2

IUPAC Name: 1-ethyl-2,3-dimethylbenzene
Synonyms: 1,2-dimethyl-3-ethylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: QUBBAXISAHIDNM-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(C)=C1C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results