accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 933727-40-3

IUPAC Name: 1-(3,4-dihydro-2H-1-benzopyran-8-yl)methanamine
Synonyms: 1-(3,4-dihydro-2H-1-benzopyran-8-yl)methanamine
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: CPJFACCHHWIMEM-UHFFFAOYSA-N
SMILES: NCC1=CC=CC2=C1OCCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results