accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOOHH3C

CAS RN 93379-49-8

IUPAC Name: [5-(hydroxydiphenylmethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]diphenylmethanol
Synonyms: [5-(hydroxydiphenylmethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]diphenylmethanol
Molecular Weight (g/mol): 466.58
Molecular Formula: C31H30O4
InChi Key: OWVIRVJQDVCGQX-UHFFFAOYNA-N
SMILES: CC1(C)OC(C(O1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results