accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 934-22-5

IUPAC Name: 1H-1,3-benzodiazol-6-amine
Synonyms: 3H-1,3-benzodiazol-5-amine
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: WFRXSXUDWCVSPI-UHFFFAOYSA-N
SMILES: NC1=CC=C2N=CNC2=C1
Quantity 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results