accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 934-72-5

IUPAC Name: 1-methanesulfinyl-4-methylbenzene
Synonyms: 1-methanesulfinyl-4-methylbenzene
Molecular Weight (g/mol): 154.23
Molecular Formula: C8H10OS
InChi Key: FEVALTJSQBFLEU-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(C)=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)