accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3C

CAS RN 934-74-7

IUPAC Name: 1-ethyl-3,5-dimethylbenzene
Synonyms: 1,3-dimethyl-5-ethylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: LMAUULKNZLEMGN-UHFFFAOYSA-N
SMILES: CCC1=CC(C)=CC(C)=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results