accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3C

CAS RN 934-80-5

IUPAC Name: 4-ethyl-1,2-dimethylbenzene
Synonyms: 4-ethyl-1,2-dimethylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: SBUYFICWQNHBCM-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C)C(C)=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results